Millicell® 24 well hanging cell culture inserts
Products
Product Category
Brand
Color
Binding Type
Diameter (mm)
Height (mm)
Surface Area cm²
Wells
Working Volume (ml)
Quality Segment
Color
Housing Material
Membrane Material
Package Size
Pore size (µm)
Sterility
Sterilization Method
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing