Skip to Content
Merck
Miconazole nitrate European Pharmacopoeia (EP) Reference Standard

Miconazole nitrate

Miconazole nitrate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(±)-Miconazole nitrate salt, 1-(2,4-Dichloro-β-[(2,4-dichlorobenzyl)oxy]phenethyl)imidazole
Empirical Formula (Hill Notation):
C18H14Cl4N2O · HNO3
CAS Number:
Molecular Weight:
479.14

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade