Methyltestosterone for system suitability
Synonym(s):
17α-Methyltestosterone, 17α-Methyl-4-androsten-17β-ol-3-one, 17β-Hydroxy-17α-methyl-4-androsten-3-one, Mesterone, Methyltestosterone
Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing