![Methyl Palmitate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/407/521/978d5732-9c91-4eca-b547-9b5c8991aa09/640/978d5732-9c91-4eca-b547-9b5c8991aa09.png)
Methyl palmitate
![Methyl Palmitate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/407/521/978d5732-9c91-4eca-b547-9b5c8991aa09/640/978d5732-9c91-4eca-b547-9b5c8991aa09.png)
Synonym(s):
n-Hexadecanoic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Methyl palmitate
Linear Formula:
CH3(CH2)14CO2CH3
CAS Number:
Molecular Weight:
270.45
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing