![Methylene Blue Trihydrate OmniPur® Grade](/deepweb/assets/sigmaaldrich/product/structures/229/061/e77dc472-7a0c-43a2-bb65-1950e95a0ad4/640/e77dc472-7a0c-43a2-bb65-1950e95a0ad4.png)
Methylene Blue Trihydrate
![Methylene Blue Trihydrate OmniPur® Grade](/deepweb/assets/sigmaaldrich/product/structures/229/061/e77dc472-7a0c-43a2-bb65-1950e95a0ad4/640/e77dc472-7a0c-43a2-bb65-1950e95a0ad4.png)
Synonym(s):
Methylene Blue trihydrate
Empirical Formula (Hill Notation):
C16H18ClN3S · 3H2O
CAS Number:
Molecular Weight:
373.90
Products
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing