Skip to Content
Merck
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3 1.4 % (w/v) in 95% ethanol

Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3

Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3 1.4 % (w/v) in 95% ethanol
Synonym(s):
Methylene Blue solution, Ehrlich’s reagent III, Loeffler’s Methylene Blue
Empirical Formula (Hill Notation):
C16H18ClN3S
CAS Number:
Molecular Weight:
319.85

Products

Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1