
Mead acid

Synonym(s):
cis-5,8,11-Eicosatrienoic acid, MEAD acid, cis,cis,cis-5,8,11-Eicosatrienoic acid
Empirical Formula (Hill Notation):
C20H34O2
CAS Number:
Molecular Weight:
306.48
Products
Brand
Formula Weight
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing