
Mangiferin

Synonym(s):
1,3,6,7-Tetrahydroxyxanthone C2-β-D-glucoside, Chinomin, Chinonin, Shamimin
Empirical Formula (Hill Notation):
C19H18O11
CAS Number:
Molecular Weight:
422.34
Products
Brand
Formula Weight
Manufacturer
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing