Skip to Content
Merck
L-N5-(1-Iminoethyl)ornithine hydrochloride ≥95% (HPLC)

L-N5-(1-Iminoethyl)ornithine hydrochloride

L-N5-(1-Iminoethyl)ornithine hydrochloride ≥95% (HPLC)
Synonym(s):
L-NIO hydrochloride, L-Ornithine ψ-acetamidine hydrochloride
Empirical Formula (Hill Notation):
C7H15N3O2 · xHCl
CAS Number:
Molecular Weight:
173.21 (free base basis)

Products

Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing