Skip to Content
Merck
L-Leucine p-nitroanilide hydrochloride ≥98% (HPLC)

L-Leucine p-nitroanilide hydrochloride

L-Leucine p-nitroanilide hydrochloride ≥98% (HPLC)
Empirical Formula (Hill Notation):
C12H17N3O3 · HCl
CAS Number:
Molecular Weight:
287.74

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing