L-Glutamic acid γ-(α-naphthylamide) monohydrate
Empirical Formula (Hill Notation):
C15H16N2O3 ·H2O
CAS Number:
Molecular Weight:
290.31
Products
Product Category
Brand
Formula Weight
Physical Form
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing