Skip to Content
Merck
L-Fenchone ≥98%

L-Fenchone

L-Fenchone ≥98%
Synonym(s):
(1R)-(−)-Fenchone, (−)-1,3,3-Trimethyl-2-norbornanone, (−)-Fenchone, (1R)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing