
KT109

Synonym(s):
(4-([1,1′-Biphenyl]-4-yl)-1H-1,2,3-triazol-1-yl)(2-benzylpiperidin-1-yl)methanone
Empirical Formula (Hill Notation):
C27H26N4O
CAS Number:
Molecular Weight:
422.52
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1