Skip to Content
Merck
Ketamine-D4 hydrochloride solution 1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®

Ketamine-D4 hydrochloride solution

Ketamine-D4 hydrochloride solution 1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C13D4H12ClNO · HCl
CAS Number:
Molecular Weight:
278.21
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique