
Isopyrazam

Synonym(s):
3-(Difluoromethyl)-1-methyl-N-[1,2,3,4-tetrahydro-9-(1-methylethyl)-1,4-methanonaphthalen-5-yl]-1H-pyrazole-4-carboxamide
Empirical Formula (Hill Notation):
C20H23F2N3O
CAS Number:
Molecular Weight:
359.41
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing