Ibuprofen impurity B
Synonym(s):
(2RS)-2-(4-Butylphenyl)propanoic acid solution, 4-Butyl-α-methylbenzeneacetic acid, p-Butylhydratropic acid
Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing