![Ibuprofen for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/644/485/da864316-5ed6-46ea-b44a-5774df5679cb/640/da864316-5ed6-46ea-b44a-5774df5679cb.png)
Ibuprofen for peak identification
![Ibuprofen for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/644/485/da864316-5ed6-46ea-b44a-5774df5679cb/640/da864316-5ed6-46ea-b44a-5774df5679cb.png)
Synonym(s):
Ibuprofen, α-Methyl-4-(isobutyl)phenylacetic acid, (±)-2-(4-Isobutylphenyl)propanoic acid
Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing