![vibrant-m](/assets/images/pdp-no-image-sq/pdp-no-image-sq_w640.png)
Hypoxanthine, monosodium salt
![vibrant-m](/assets/images/pdp-no-image-sq/pdp-no-image-sq_w640.png)
Synonym(s):
Hypoxanthine monosodium salt
Empirical Formula (Hill Notation):
C5H3N4O · Na
Molecular Weight:
158.09
Products
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing