
HU-210

Synonym(s):
(6aR-trans-3-(1, 1-Dimethylheptyl)-6a,7,10,10a-tetrahydro-1-hydroxy-6,6-dimethyl-6H-dibenzo[b,d]pyran-9-methanol
Empirical Formula (Hill Notation):
C25H38O3
CAS Number:
Molecular Weight:
386.57
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing