HEPES sodium salt solution
Empirical Formula (Hill Notation):
C8H17N2NaO4S
CAS Number:
Molecular Weight:
260.29
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Quality Segment
Special Grade
Sterilization Method
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing