Skip to Content
Merck
Guanabenz acetate salt powder

Guanabenz acetate salt

Guanabenz acetate salt powder
Synonym(s):
1-(2,6-Dichlorobenzylideneamino)guanidine acetate salt, WY-8678
Empirical Formula (Hill Notation):
C8H8Cl2N4 · C2H4O2
CAS Number:
Molecular Weight:
291.13
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing