Skip to Content
Merck
γ-Octalactone natural, ≥97%, FG

γ-Octalactone

γ-Octalactone natural, ≥97%, FG
Synonym(s):
γ-Octanoic lactone, 4-Hydroxy octanoic acid, Octano-1,4-lactone
Empirical Formula (Hill Notation):
C8H14O2
CAS Number:
Molecular Weight:
142.20
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing