Skip to Content
Merck
[Glu1]-Fibrinopeptide B human ≥90% (HPLC)

[Glu1]-Fibrinopeptide B human

[Glu1]-Fibrinopeptide B human ≥90% (HPLC)
Empirical Formula (Hill Notation):
C66H95N19O26
CAS Number:
Molecular Weight:
1570.57

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing