![Fluorouracil impurity F European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/254/129/12ed70f3-ac8f-4041-bb21-a096e9bb5781/640/12ed70f3-ac8f-4041-bb21-a096e9bb5781.png)
Fluorouracil impurity F
![Fluorouracil impurity F European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/254/129/12ed70f3-ac8f-4041-bb21-a096e9bb5781/640/12ed70f3-ac8f-4041-bb21-a096e9bb5781.png)
Synonym(s):
5-Fluoro-2-ethoxy-4(1H)pyrimidinone, 2-Ethoxy-5-fluoropyrimidin-4(1H)-one
Linear Formula:
C4N2H2FOOCH2CH3
CAS Number:
Molecular Weight:
158.13
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing