
Fibroblast Growth Factor-Basic, human

Synonym(s):
Fibroblast Growth Factor-Basic human, FGF2, bFGF
CAS Number:
Products
Product Category
Brand
Biological Source
Color
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing