Fexofenadine methyl ester
Synonym(s):
Methyl 2-[4-[1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl]phenyl]-2-methylpropanoate
Empirical Formula (Hill Notation):
C33H41NO4
CAS Number:
Molecular Weight:
515.68
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing