Skip to Content
Merck
Famciclovir Related Compound E certified reference material, pharmaceutical secondary standard

Famciclovir Related Compound E

Famciclovir Related Compound E certified reference material, pharmaceutical secondary standard
Synonym(s):
2-Aminopurine
Empirical Formula (Hill Notation):
C5H5N5
CAS Number:
Molecular Weight:
135.13

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1