Skip to Content
Merck
Erythrosin B Dye content ≥95 %

Erythrosin B

Erythrosin B Dye content ≥95 %
Synonym(s):
Solvent Red 140
Empirical Formula (Hill Notation):
C20H8I4O5
CAS Number:
Molecular Weight:
835.89
EC Number:

Products

Product Category
Color
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing