![Endrin ketone PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/280/965/57282c58-94e8-4fbc-bcaa-131bdd257147/640/57282c58-94e8-4fbc-bcaa-131bdd257147.png)
Endrin ketone
![Endrin ketone PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/280/965/57282c58-94e8-4fbc-bcaa-131bdd257147/640/57282c58-94e8-4fbc-bcaa-131bdd257147.png)
Empirical Formula (Hill Notation):
C12H8Cl6O
CAS Number:
Molecular Weight:
380.91
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing