Skip to Content
Merck
Endoglycosidase F2 from Elizabethkingia miricola recombinant, expressed in E. coli, 20 U/mg

Endoglycosidase F2 from Elizabethkingia miricola

Endoglycosidase F2 from Elizabethkingia miricola recombinant, expressed in E. coli, 20 U/mg
Synonym(s):
Elizabethkingia miricola, Endo-β-N-acetylglucosaminidase F2, Endo F2, Endoglycosidase F2 from Chryseobacterium meningosepticum, Endoglycosidase F2 from Elizabethkingia meningoseptica, Endoglycosidase F2 from Flavobacterium meningosepticum
CAS Number:

Products

Product Category
Brand
Physical Form
Recombinant Host
Quality Segment
Showing 1-1 of 1