
DL-Dithiothreitol-d10

Synonym(s):
Cleland’s reagent-d10
Linear Formula:
DSCD2CD(OD)CD(OD)CD2SD
CAS Number:
Molecular Weight:
164.31
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing