Diosmin for system suitability
Synonym(s):
Diosmin, 3′,5,7-Trihydroxy-4′-methoxyflavone 7-rutinoside, Barosmin, Buchu resin, Diosmetin 7-O-rhamnosylglucoside, Diosmetin 7-rutinoside
Empirical Formula (Hill Notation):
C28H32O15
CAS Number:
Molecular Weight:
608.54
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing