Skip to Content
ID
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diosmetin
Synonym(s):
3′,5,7-Trihydroxy-4′-methoxyflavone, 4′-Methylluteolin, 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chromen-4-one, Luteolin 4′-methyl ether
Empirical Formula (Hill Notation):
C
16
H
12
O
6
CAS Number:
520-34-3
Molecular Weight:
300.26
EC Number:
208-291-8
Products
Product Category
antioxidants (1)
flavonoids (1)
metabolites (1)
natural products (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Physical Form
Purity
Research Area
Application
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
D7321
Diosmetin,
Expand
View Pricing