
Diethylene glycol monomethyl ether

Synonym(s):
Diethylene glycol monomethyl ether
Empirical Formula (Hill Notation):
C5H12O3
CAS Number:
Molecular Weight:
120.15
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing