![Diclazuril for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/122/500/ec845083-54e8-4de2-882f-f1255dff7efb/640/ec845083-54e8-4de2-882f-f1255dff7efb.png)
Diclazuril for system suitability
![Diclazuril for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/122/500/ec845083-54e8-4de2-882f-f1255dff7efb/640/ec845083-54e8-4de2-882f-f1255dff7efb.png)
Synonym(s):
2,6-Dichloro-α-(4-chlorophenyl)-4-[4,5-dihydro-3,5-dioxo-1,2,4-triazin-2(3H)-yl]benzeneacetonitrile
Empirical Formula (Hill Notation):
C17H9Cl3N4O2
CAS Number:
Molecular Weight:
407.64
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing