Skip to Content
ID
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dibenzo[
a
,
l
]pyrene
Empirical Formula (Hill Notation):
C
24
H
14
CAS Number:
191-30-0
Molecular Weight:
302.37
EC Number:
205-886-4
Products
Brand
Supelco (1)
Formula Weight
Manufacturer
Agency
Format
Application
Package Size
Reference Material Type
Special Grade
certified reference material (2)
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
BCR096
Dibenzo[
a
,
l
]pyrene,
BCR
®
, certified reference material
Expand
View Pricing
ERD-051
Dibenzo[
a
,
l
]pyrene,
vial of 25 mg, Cerilliant
®
Expand
View Pricing