di-Ammonium hydrogen phosphate
Synonym(s):
di-Ammonium hydrogen phosphate
Linear Formula:
(NH4)2HPO4
CAS Number:
Molecular Weight:
132.06
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing