Skip to Content
Merck
Decanoyl-L-carnitine analytical standard

Decanoyl-L-carnitine

Decanoyl-L-carnitine analytical standard
Synonym(s):
(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-1-propanaminium inner salt, L-Carnitine decanoyl ester, C10-Carnitine
Empirical Formula (Hill Notation):
C17H33NO4
CAS Number:
Molecular Weight:
315.45

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing