Skip to Content
Merck
D-(+)-Melezitose monohydrate suitable for microbiology, ≥99.0%

D-(+)-Melezitose monohydrate

D-(+)-Melezitose monohydrate suitable for microbiology, ≥99.0%
Synonym(s):
O-α-D-Glucopyranosyl-(1→3)-β-D-fructofuranosyl-α-D-glucopyranoside
Empirical Formula (Hill Notation):
C18H32O16 · H2O
CAS Number:
Molecular Weight:
522.45
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing