D(−)-2-Amino-5-phosphonopentanoic acid
Synonym(s):
D(−)-AP-5, D(−)-APV, D-2-Amino-5-phosphonovaleric acid
Empirical Formula (Hill Notation):
C5H12NO5P
CAS Number:
Molecular Weight:
197.13
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing