Skip to Content
Merck
Cyphenothrin PESTANAL®, analytical standard, sum of isomers

Cyphenothrin

Cyphenothrin PESTANAL®, analytical standard, sum of isomers
Empirical Formula (Hill Notation):
C24H25NO3
CAS Number:
Molecular Weight:
375.46
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing