
Cyclomethicone 5

Synonym(s):
Decamethylcyclopentasiloxane
Empirical Formula (Hill Notation):
C10H30O5Si5
CAS Number:
Molecular Weight:
370.77
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing