![Corosolic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/406/793/36df9e7e-4dd3-4112-a187-3abb2dd710d5/640/36df9e7e-4dd3-4112-a187-3abb2dd710d5.png)
Corosolic acid
![Corosolic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/406/793/36df9e7e-4dd3-4112-a187-3abb2dd710d5/640/36df9e7e-4dd3-4112-a187-3abb2dd710d5.png)
Synonym(s):
(2α,3β)-2,3-Dihydroxyurs-12-en-28-oic acid, 2α-Hydroxyursolic acid, Colosolic acid, Corsolic acid, Glucosol
Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70
Products
Brand
Formula Weight
Manufacturer
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing