Skip to Content
Merck
(+/-)-Cloprostenol sodium salt hydrate ≥98% (HPLC), powder

(+/-)-Cloprostenol sodium salt hydrate

(+/-)-Cloprostenol sodium salt hydrate ≥98% (HPLC), powder
Synonym(s):
(+/-)-16-(3-Chlorophenoxy)-9,11,15R-trihydroxy-17,18,19,20-tetranorprosta-5Z,13E-dien-1-oic acid sodium salt hydrate
Empirical Formula (Hill Notation):
C22H28ClNaO6 · xH2O
CAS Number:
Molecular Weight:
446.90 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing