![Clopidogrel for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/198/766/899d94cf-b497-48d6-9cd9-4716fb41ac49/640/899d94cf-b497-48d6-9cd9-4716fb41ac49.png)
Clopidogrel for system suitability
![Clopidogrel for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/198/766/899d94cf-b497-48d6-9cd9-4716fb41ac49/640/899d94cf-b497-48d6-9cd9-4716fb41ac49.png)
Synonym(s):
(S)-Methyl (2-chlorophenyl)(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate hydrogensulfate
Empirical Formula (Hill Notation):
C16H16ClNO2S · H2SO4
CAS Number:
Molecular Weight:
419.90
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing