![Cefalexin British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/128/336/68b016ea-6906-4737-ad49-eee672629615/640/68b016ea-6906-4737-ad49-eee672629615.png)
Cefalexin
![Cefalexin British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/128/336/68b016ea-6906-4737-ad49-eee672629615/640/68b016ea-6906-4737-ad49-eee672629615.png)
Synonym(s):
7-(D-α-amino-phenylacetamido)-3-methyl-3-cepheme-4-carboxylic acid
Empirical Formula (Hill Notation):
C16H17N3O4S
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing