Carbotrap® 300 Thermal Desorption Tube
Synonym(s):
Carbotrap® 300
Products
Product Category
Brand
Sustainability
Feature
Product Line
Agency
ID (mm)
Length (in)
OD (mm)
Compatibility
Application
Package Size
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing