Skip to Content
Merck
Butyl (S)-(−)-lactate ≥97%, FG

Butyl (S)-(−)-lactate

Butyl (S)-(−)-lactate ≥97%, FG
Synonym(s):
(−)-Butyl L-lactate
Linear Formula:
CH3CH(OH)CO2(CH2)3CH3
CAS Number:
Molecular Weight:
146.18

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing