Skip to Content
Merck
Butan-3-one-2-yl butanoate ≥98%, FCC, FG

Butan-3-one-2-yl butanoate

Butan-3-one-2-yl butanoate ≥98%, FCC, FG
Linear Formula:
CH3CH2CH2CO2CH(CH3)COCH3
CAS Number:
Molecular Weight:
158.19
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing