Skip to Content
Merck
BIOshell Glycan HPLC Columns L × I.D. 15 cm × 2.1 mm, 2.7 μm particle size

BIOshell Glycan (2.7 μm) HPLC Columns

BIOshell Glycan HPLC Columns L × I.D. 15 cm × 2.1 mm, 2.7 μm particle size

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Analyte Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1